Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Arylsulfatase G/ARSG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4600SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4600SP 25 μg
AF4600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4600SP Supplier R&D Systems Supplier No. AF4600SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Arylsulfatase G/ARSG Polyclonal specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Arylsulfatase G/ARSG
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q3TYD4
Gene Alias ARSG, arylsulfatase G, ASG, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG
Gene Symbols ARSG
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Arylsulfatase G/ARSG Gly17-Val525 Accession # Q3TYD4
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22901
Test Specificity Detects mouse Arylsulfatase G/ARSG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse ARSA, recombinant human (rh) ARSB and rhARSJ is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.