Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4886SP
This item is not returnable.
View return policy
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Polyclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, NAAA, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, Nacylethanolaminehydrolyzing Acid Amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
Goat | |
Affinity Purified | |
RUO | |
27163 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
AAH04572 | |
NAAA | |
Mouse myeloma cell line NS0-derived recombinant mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A Val33-Ser362 Accession # AAH04572 | |
25 μg | |
Primary | |
Detects mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ASAHL is observed and less than 5% cross-reactivity rhASAHL-2 and recombinant mouse ASAHL-2 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction