Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4886 Supplier R&D Systems Supplier No. AF4886
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Polyclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. AAH04572
Gene Alias Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, NAAA, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, Nacylethanolaminehydrolyzing Acid Amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT
Gene Symbols NAAA
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A Val33-Ser362 Accession # AAH04572
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27163
Test Specificity Detects mouse ASAHL/N-acylethanolamine-hydrolyzing Acid A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ASAHL is observed and less than 5% cross-reactivity rhASAHL-2 and recombinant mouse ASAHL-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.