Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ASGR1/ASGPR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB27552SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB27552SP 25 μg
MAB27552100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB27552SP Supplier R&D Systems Supplier No. MAB27552SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

ASGPR1 Monoclonal specifically detects ASGPR1 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen ASGPR1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 352803
Conjugate Unconjugated
Dilution Western Blot 0.5 μg;/mL, ELISA, Immunohistochemistry 5-25 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_033844
Gene Alias ASGPR, ASGPR 1, ASGP-R 1, ASGPR1, asialoglycoprotein receptor 1, CLEC4H1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1, MHL1, RHL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ASGR1/ASGPR1, Ser60-Asn284, Accession No. NP_033844
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 432.0
Test Specificity Detects mouse ASGR1/ASGPR1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.