Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Autoimmune Regulator/AIRE Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6184N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6184N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6184N100 Supplier R&D Systems Supplier No. IC6184N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Autoimmune Regulator/AIRE Monoclonal specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Autoimmune Regulator/AIRE
Applications Flow Cytometry
Classification Monoclonal
Clone 609930
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9Z0E3
Gene Alias AIRE, APECED, APECED protein, APECEDAPSI, APS1, APSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1
Gene Symbols AIRE
Host Species Rat
Immunogen E. coli-derived recombinant mouse AIRE Ser476-Ser552 (predicted) Accession # Q9Z0E3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326
Test Specificity Detects mouse AIRE in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human AIRE is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.