Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Axl Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB8541U100UG
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Specifications
Axl | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q00993 | |
AXL | |
Mouse myeloma cell line NS0-derived recombinant mouse Axl Ala19-Trp445 Accession # Q00993 | |
RUO | |
558 | |
Mouse | |
IgG2a |
Flow Cytometry | |
175128 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
Rat | |
100 μg | |
Primary | |
Detects mouse Axl. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction