Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Axl PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems FAB8541P
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Specifications
Axl | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
558 | |
Mouse | |
Purified |
Flow Cytometry | |
175128 | |
Flow Cytometry 10 uL/10^6 cells | |
Q00993 | |
AXL | |
Mouse myeloma cell line NS0-derived recombinant mouse Axl Ala19-Trp445 Accession # Q00993 | |
100 Tests | |
Primary | |
Detects mouse Axl. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction