Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-1/CD80 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF740
This item is not returnable.
View return policy
Description
B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
B7-1/CD80 | |
Polyclonal | |
Unconjugated | |
Q00609 | |
CD80 | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-1/CD80 Asp37-Lys245 Accession # Q00609 | |
100 μg | |
Primary | |
Detects mouse B7-1/CD80 in ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF, ELISA | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.15-0.6 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Goat | |
Affinity Purified | |
RUO | |
941 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction