Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-H2 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB158U100UG
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry.Specifications
B7-H2/ICOSLG | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9JHJ8 | |
Icosl | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 Glu47-Lys279 Accession # Q9JHJ8 | |
RUO | |
23308 | |
Mouse | |
IgG1 |
Flow Cytometry | |
599841 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Rat | |
100 μg | |
Primary | |
Detects mouse B7-H2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-H2, rhB7-H3, rhB7-H3b, rhPD-L2, recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H3, rmB7-H4, rmPD-L2, or recombinant rat B7-2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction