Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-H2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | B7-H2/ICOSLG |
---|---|
Clone | 987602 |
Dilution | Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction 0.5 μg/106 cells, Neutralization, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, Neutralization, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10083SP
![]() |
R&D Systems
MAB10083SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10083100
![]() |
R&D Systems
MAB10083100 |
100 μg |
Each for $467.50
|
|
|||||
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Neutralization, CyTOF-ready.Specifications
B7-H2/ICOSLG | |
Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction 0.5 μg/106 cells, Neutralization, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
Q9JHJ8 | |
23308 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
987602 | |
Flow Cytometry, Blocking Assay, Neutralization, CyTOF | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H2, Met1-Lys279, Accession # Q9JHJ8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse B7-H2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title