Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-H3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$162.90 - $502.50
Specifications
Antigen | B7-H3/CD276 |
---|---|
Clone | 2741D |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1397SP
![]() |
R&D Systems
MAB1397SP |
25 μg |
Each for $162.90
|
|
|||||
MAB1397100
![]() |
R&D Systems
MAB1397100 |
100 μg |
Each for $502.50
|
|
|||||
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Mouse samples. It is validated for Western BlotSpecifications
B7-H3/CD276 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Mouse myeloma cell line NS0-derived mouse B7-H3, Val29-Phe244, Accession # Q8VE98.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2741D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Q8VE98.1 | |
80381 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title