Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-H3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1397
This item is not returnable.
View return policy
Description
B7-H3/CD276 Polyclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for Western Blot.Specifications
B7-H3/CD276 | |
Polyclonal | |
Biotin | |
Q8VE98 | |
CD276 | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H3 Val29-Phe244 Accession # Q8VE98 | |
50 μg | |
Primary | |
Detects mouse B7-H3 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human B7-H3 is observed and less than 1% cross-reactivity with recombinant mouse (rm) B7-H2, rmB7-H1, and rmB7-1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Goat | |
Affinity Purified | |
RUO | |
80381 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction