Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse B7-H4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2154
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2154 100 μg
MAB2154SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2154 Supplier R&D Systems Supplier No. MAB2154
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen B7-H4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 297219
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7TSP5
Gene Alias B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1
Gene Symbols VTCN1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse B7-H4 Phe29-Pro258 Accession # Q7TSP5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79679
Test Specificity Detects mouse B7-H4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H2, rmB7-H3, recombinant human (rh) B7-H3b, rhB7-H4, or rmPD-L2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.