Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BACH1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5777
This item is not returnable.
View return policy
Description
BACH1 Polyclonal specifically detects BACH1 in Mouse samples. It is validated for Western Blot.Specifications
BACH1 | |
Polyclonal | |
Unconjugated | |
P97302 | |
BACH1 | |
E. coli-derived recombinant mouse BACH1 Ser133-Gly513 Accession # P97302 | |
100 μg | |
Primary | |
Detects endogenous mouse BACH1 in Western blots. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
basic region leucine zipper transcriptional regulator BACH1, BTB, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1 | |
Goat | |
Affinity Purified | |
RUO | |
571 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction