Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BAFF/BLyS/TNFSF13B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1357500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1357500 500 μg
MAB1357SP 25 μg
MAB1357100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1357500 Supplier R&D Systems Supplier No. MAB1357500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 9 publications

BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen BAFF/BLyS/TNFSF13B
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 121808
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9WU72
Gene Alias ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Gene Symbols TNFSF13B
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BAFF Ala127-Leu309 Accession # Q9WU72
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Test Specificity Detects mouse BAFF in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) 4-1BB Ligand, rmEDA, rmFas Ligand, rmOX40 Ligand, rmTNF-alpha , rmTRANCE, rmTWEAK, recombinant human (rh) APRIL, rhBAFF, rhEDA-A2, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1/beta 2, rhLymphotoxin alpha 2/beta 1, rhTNF-alpha , rhTRAIL, rhVEGI, recombinant canine, cotton rat, equine, feline, porcine, or rat TNF-alpha .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.