Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BAFF/BLyS/TNFSF13B Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | BAFF/BLyS/TNFSF13B |
---|---|
Clone | 121834 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13571SP
![]() |
R&D Systems
MAB13571SP |
25 μg |
Each for $200.50
|
|
|||||
MAB13571100
![]() |
R&D Systems
MAB13571100 |
100 μg |
Each for $456.00
|
|
|||||
Description
BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Mouse samples. It is validated for Immunocytochemistry.Specifications
BAFF/BLyS/TNFSF13B | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 | |
Mouse myeloma cell line NS0-derived recombinant mouse BAFF/BLyS/TNFSF13B, Ala127-Leu309, Accession # Q9WU72 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse BAFF/BLyS/TNFSF13B in direct ELISAs. |
121834 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Q9WU72 | |
10673 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title