Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BAMBI/NMA Biotinylated Antibody, R&D Systems™

Catalog No. BAF2387
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2387 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2387 Supplier R&D Systems Supplier No. BAF2387
This item is not returnable. View return policy

Goat Polyclonal Antibody

BAMBI/NMA Polyclonal specifically detects BAMBI/NMA in Mouse samples. It is validated for Western Blot.

Specifications

Antigen BAMBI/NMA
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9D0L6
Gene Alias BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMA, NMANon-metastatic gene A protein, Putative transmembrane protein NMA
Gene Symbols BAMBI
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BAMBI/NMA Ile28-Thr143 Accession # Q9D0L6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25805
Test Specificity Detects mouse BAMBI/NMA in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.