Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BCMA/TNFRSF17 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB593
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
BCMA/TNFRSF17 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
608 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Blockade of Receptor-ligand Interaction | |
161616 | |
Blockade of Receptor-ligand Interaction | |
O88472 | |
TNFRSF17 | |
Mouse myeloma cell line NS0-derived recombinant mouse BCMA Met1-Thr49 Accession # O88472 | |
500 μg | |
Primary | |
Detects mouse BCMA in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) BCMA or rhTACI. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction