Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BCMA/TNFRSF17 Fluorescein-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems FAB593F
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Mouse samples. It is validated for Flow Cytometry.Specifications
BCMA/TNFRSF17 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
608 | |
Mouse | |
Purified |
Flow Cytometry | |
161616 | |
Flow Cytometry 10 uL/10^6 cells | |
O88472 | |
TNFRSF17 | |
Mouse myeloma cell line NS0-derived recombinant mouse BCMA Met1-Thr49 Accession # O88472 | |
100 Tests | |
Primary | |
Detects mouse BCMA in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) BCMA or rhTACI. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction