Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse beta 2-Microglobulin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB8325
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Mouse samples. It is validated for Western Blot.Specifications
beta 2-Microglobulin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B2M, beta 2Microglobulin, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
567 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
893803 | |
Western Blot 0.5 ug/mL | |
P01887 | |
B2M | |
Mouse myeloma cell line NS0-derived recombinant mouse beta 2 -Microglobulin Met1-Met119 Accession # P01887 | |
100 μg | |
Primary | |
Detects mouse beta 2 -Microglobulin in direct ELISAs and Western blots. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction