Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse beta IG-H3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2559
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2559 100 μg
MAB2559SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2559 Supplier R&D Systems Supplier No. MAB2559
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Beta Ig-h3/TGFBI Monoclonal specifically detects Beta Ig-h3/TGFBI in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Beta Ig-h3/TGFBI
Applications Western Blot
Classification Monoclonal
Clone 320305
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P82198
Gene Alias beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3
Gene Symbols TGFBI
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse beta IG-H3 Gly24-His683 Accession # P82198
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7045
Test Specificity Detects mouse beta IG-H3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human beta IG-H3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.