Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse beta-NGF Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB11561SP
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal specifically detects beta-NGF in Mouse samples. It is validated for ELISA.Specifications
beta-NGF | |
Monoclonal | |
Unconjugated | |
Q6LDU8 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
4803 | |
Mouse | |
Purified |
ELISA | |
2529A | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Beta nerve growth factor precursor, beta-nerve growth factor, betaNGF, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFB, NGFBMGC161428 | |
Mouse myeloma cell line, NS0-derived mouse beta-NGF, Ser122-Gly241, Accession # Q6LDU8 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction