Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Betacellulin/BTC Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1025
This item is not returnable.
View return policy
Description
Betacellulin/BTC Polyclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
Betacellulin/BTC | |
Polyclonal | |
Biotin | |
Q543J8 | |
BTC | |
E. coli-derived recombinant mouse Betacellulin/BTC Asp32-Gln118 Accession # Q543J8 | |
50 μg | |
Primary | |
Detects mouse Betacellulin/BTC in ELISAs and Western blots. In sandwich immunoassays, less than 0.02% cross-reactivity with recombinant mouse (rm) ErbB4, rmEGF R, rmAR, recombinant human (rh) BTC, rhTGF-alpha , and rhHB-EGF is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
betacellulin, BTC, probetacellulin | |
Goat | |
Affinity Purified | |
RUO | |
685 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction