Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse BID ELISA Kit (Colorimetric)
SDP

Catalog No. NBP269967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP269967 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP269967 Supplier Novus Biologicals™ Supplier No. NBP269967
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse BID ELISA Kit (Colorimetric) measures mouse BID in serum, plasma and other biological fluids.

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Cross Reactivity This kit recognizes Mouse BID in samples. No significant cross-reactivity or interference between Mouse BID and analogues was observed.
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Plasma, Serum, Serum
Specificity This kit recognizes Mouse BID in samples. No significant cross-reactivity or interference between Mouse BID and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 637
Gene Symbol BID
Host Species Mouse
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1 b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid
Target BID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.