Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BMP-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6038SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6038SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6038SP Supplier R&D Systems Supplier No. MAB6038SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

BMP-10 Monoclonal specifically detects BMP-10 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen BMP-10
Applications Immunohistochemistry
Classification Monoclonal
Clone 654319
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9R229
Gene Alias BMP10, BMP-10, bone morphogenetic protein 10, MGC126783
Gene Symbols BMP10
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human BMP-10 Asn314-Arg421 Accession # Q9R229
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27302
Test Specificity Detects mouse BMP-10 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) BMP-10 and less than 10% cross-reactivity with rhBMP-8a and recombinant mouse (rm) BMP-8b is observed. No cross-reactivity with rmBMP-3, -4, -5, -6, -7, -9, -15, rhBMP-1.1, -2, -3b, or the propeptide of rhBMP-10 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.