Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BMP-6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6325SP
This item is not returnable.
View return policy
Description
BMP-6 Polyclonal specifically detects BMP-6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BMP-6 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, Vgr-1, VGR1 | |
Sheep | |
Affinity Purified | |
RUO | |
654 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P20722 | |
BMP6 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse BMP-6 Ser372-His510 Accession # P20722 | |
25 μg | |
Primary | |
Detects recombinant human and mouse BMP-6 in direct ELISAs and detects mouse BMP-6 in Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) BMP-5 and rmBMP-7 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction