Learn More
Novus Biologicals™ Mouse Brain Whole Tissue Lysate (Adult Whole Normal)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Quantity | 0.5 mL |
| Storage Buffer | 1X SDS-PAGE Sample Buffer (62.5 mM Tris HCl, 2% SDS, 10% Glycerol and 0.005% bromophenol blue, pH 6.8) 10% (v/v) Glycerol |
| Purity | Multi-step |
| Test Specificity | Normal Mouse Brain whole cell extract lysate |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Host Species | Mouse |
| Concentration | 1 mg/mL |
| Product Type | Mouse Brain Whole Tissue Lysate (Adult Normal) |
| Content And Storage | Store at -70°C. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.