Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse BTLA Alexa Fluor 488-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems FAB3007G
This item is not returnable.
View return policy
Description
BTLA/CD272 Polyclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Flow Cytometry.Specifications
BTLA/CD272 | |
Polyclonal | |
Flow Cytometry 5 uL/10^6 cells | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Goat | |
Affinity Purified | |
RUO | |
151888 | |
Mouse | |
IgG |
Flow Cytometry | |
Alexa Fluor 488 | |
Q32MV9 | |
BTLA | |
Mouse myeloma cell line NS0-derived recombinant mouse BTLA Glu30-Gly176 (Pro41Glu, Thr45Asn, Thr47Lys, Gln52His, Arg55Trp, Gln63Glu, Cys85Trp) Accession # Q32MV9 | |
100 Tests | |
Primary | |
Detects mouse BTLA in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human BTLA is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction