Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BTN1A1/Butyrophilin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4765
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4765 Supplier R&D Systems Supplier No. AF4765
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BTN1A1/Butyrophilin Polyclonal specifically detects BTN1A1/Butyrophilin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen BTN1A1/Butyrophilin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Gene Accession No. Q62556
Gene Alias BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Gene Symbols BTN1A1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BTN1A1/Butyrophilin Ala27-Trp247 Accession # Q62556
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696
Test Specificity Detects mouse BTN1A1/Butyrophilin in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human BTN1A1/Butyrophilin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.