Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse C1qR1/CD93 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1696S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1696S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1696S100 Supplier R&D Systems Supplier No. FAB1696S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

C1qR1/CD93 Monoclonal specifically detects C1qR1/CD93 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen C1qR1/CD93
Applications Flow Cytometry
Classification Monoclonal
Clone 223437
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O89103
Gene Alias AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Gene Symbols CD93
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse C1q R1/CD93 Ala23-Asn572 Accession # O89103
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22918
Test Specificity Detects mouse C1q R1/CD93 in direct ELISAs.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.