Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse C1qR1/CD93 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1696
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1696 100 μg
AF1696SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1696 Supplier R&D Systems Supplier No. AF1696
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 3 publications

C1qR1/CD93 Polyclonal specifically detects C1qR1/CD93 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen C1qR1/CD93
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O89103
Gene Alias AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Gene Symbols CD93
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse C1q R1/CD93 Ala23-Asn572 Accession # O89103
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22918
Test Specificity Detects mouse C1q R1/CD93 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.