Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse C4.4A/LYPD3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5567
This item is not returnable.
View return policy
Description
C4.4A/LYPD3 Polyclonal specifically detects C4.4A/LYPD3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
C4.4A/LYPD3 | |
Polyclonal | |
Unconjugated | |
Q91YK8 | |
LYPD3 | |
Mouse myeloma cell line NS0-derived recombinant mouse C4.4A Leu33-His287 Accession # Q91YK8 | |
100 μg | |
Primary | |
Detects mouse C4.4A in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human C4.4A is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, LYPD3, Matrigel-induced gene C4 protein, MIG-C4 | |
Goat | |
Affinity Purified | |
RUO | |
27076 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction