Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Carbohydrate Sulfotransferase 7/CHST7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5108
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5108 100 μg
AF5108SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF5108 Supplier R&D Systems Supplier No. AF5108
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carbohydrate Sulfotransferase 7/CHST7 Polyclonal specifically detects Carbohydrate Sulfotransferase 7/CHST7 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Carbohydrate Sulfotransferase 7/CHST7
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q9EP78
Gene Alias C6ST2, carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7, CHST7, GlcNAc6ST-4, GST-5
Gene Symbols CHST7
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CHST7 Ser34-Val484 Accession # Q9EP78
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56548
Test Specificity Detects mouse Sulfotransferase 7/CHST7 in direct ELISAs and Western blots. In direct ELISA, less than 1% cross-reactivity with recombinant mouse (rm) CHST1, recombinant human CHST2, rmCHST5, and rmCHST3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.