Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Carbonic Anhydrase IX/CA9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2344SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2344SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2344SP Supplier R&D Systems Supplier No. AF2344SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 12 publications

Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8VHB5
Gene Alias CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Gene Symbols Car9
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase IX/CA9 Gln32-Asp390 Accession # Q8VHB5
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 768
Test Specificity Detects mouse Carbonic Anhydrase IX/CA9 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) CA9 is observed, 5% cross-reactivity with recombinant mouse (rm) CA14 is observed, and less than 1% cross-reactivity with rhCA1, -2, -3, -8, -10, -13, rmCA4, and -12 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.