Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2345SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2345SP 25 μg
MAB2345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2345SP Supplier R&D Systems Supplier No. MAB2345SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Carbonic Anhydrase XII/CA12 Monoclonal specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Carbonic Anhydrase XII/CA12
Applications Western Blot
Classification Monoclonal
Clone 336103
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8CI85
Gene Alias CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Gene Symbols Car12
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12 Ala25-Ser301 Accession # Q8CI85
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 771
Test Specificity Detects mouse Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) CA12, 10% cross-reactivity with rhCA2, and no cross-reactivity with rhCA1, 3, 5A, 5B, 6, 7, 8, 10, 11, 13, recombinant mouse CA4, 9, or 14 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.