Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Carboxylesterase 2/CES2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5280
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5280 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5280 Supplier R&D Systems Supplier No. AF5280
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carboxylesterase 2/CES2 Polyclonal specifically detects Carboxylesterase 2/CES2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Carboxylesterase 2/CES2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q91WG0
Gene Alias carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2
Gene Symbols Ces2c
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 2/CES2 Gln27-Lys557 (predicted) Accession # Q91WG0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8824
Test Specificity Detects mouse Carboxylesterase 2/CES2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CES2 is observed and approximately 5% cross-reactivity with recombinant mouse (rm) CES3 and rmCES5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.