Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Carboxypeptidase A1/CPA1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF2765
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Polyclonal specifically detects Carboxypeptidase A1/CPA1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Carboxypeptidase A1/CPA1 | |
Polyclonal | |
Unconjugated | |
Q7TPZ8 | |
CPA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase A1/CPA1 Asn17-Tyr419 Accession # Q7TPZ8 | |
100 μg | |
Primary | |
Detects mouse Carboxypeptidase A1/CPA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) CPA1 is observed and approximately 10% cross-reactivity with rhCPA2, rhCPB1, and recombinant mouse CPA4 is observed. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2 ug/mL | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, CPA1, EC 3.4.17, EC 3.4.17.1 | |
Goat | |
Affinity Purified | |
RUO | |
1357 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction