Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Carboxypeptidase B1/CPB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2898
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2898 100 μg
AF2898SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2898 Supplier R&D Systems Supplier No. AF2898
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carboxypeptidase B1/CPB1 Polyclonal specifically detects Carboxypeptidase B1/CPB1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Carboxypeptidase B1/CPB1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. NP_083982
Gene Alias carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Gene Symbols CPB1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase B1/CPB1 Ser16-Tyr415 Accession # NP_083982
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1360
Test Specificity Detects mouse Carboxypeptidase B1/CPB1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPB1 is observed and less than 1% cross-reactivity with rhCPA1 and recombinant mouse CPA4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.