Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Carboxypeptidase X1/CPXM1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2785
This item is not returnable.
View return policy
Description
Carboxypeptidase X1/CPXM1 Polyclonal specifically detects Carboxypeptidase X1/CPXM1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase X1/CPXM1 | |
Polyclonal | |
Unconjugated | |
Q9Z100 | |
CPXM1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase X1/CPXM1 Ala21-Lys722 Accession # Q9Z100 | |
100 μg | |
Primary | |
Detects mouse Carboxypeptidase X1/CPXM1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human CPXM1 is observed. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
carboxypeptidase X (M14 family), carboxypeptidase X (M14 family), member 1, CPX1, CPX-1, CPX1EC 3.4.17.-, CPXM, CPXM1, EC 3.4.17, EC 3.4.17.10, EC 3.4.17.3, Metallocarboxypeptidase CPX-1, probable carboxypeptidase X1 | |
Goat | |
Affinity Purified | |
RUO | |
56265 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction