Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Cardiotrophin-1/CT-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB438SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB438SP 25 μg
MAB438 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB438SP Supplier R&D Systems Supplier No. MAB438SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Cardiotrophin-1/CT-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 91907
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q60753
Gene Alias cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1
Gene Symbols CTF1
Host Species Rat
Immunogen E. coli-derived recombinant mouse Cardiotrophin-1 Ser2-Ala203 Accession # Q60753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489
Test Specificity Detects mouse Cardiotrophin-1 in ELISAs and Western blots. In ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Cardiotrophin-1 and no cross-reactivity with rmIL-11, rmIL-6, rrCNTF, rmLIF, or rmOSM.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.