Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cathepsin A/Lysosomal Carboxypeptidase A Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB11231SP
This item is not returnable.
View return policy
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation.Specifications
Cathepsin A/Lysosomal Carboxypeptidase A | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5476 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Immunohistochemistry, Immunoprecipitation | |
191424 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CTSA | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin A aa 24-474 | |
25 μg | |
Primary | |
Detects mouse Cathepsin A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Cathepsin A or recombinant mouse DPPIV/CD26 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction