Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cathepsin D Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1029
This item is not returnable.
View return policy
Description
Cathepsin D Polyclonal specifically detects Cathepsin D in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cathepsin D | |
Polyclonal | |
Biotin | |
Q3UCD9 | |
CTSD | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin D Ile21-Leu410 Accession # Q3UCD9 | |
50 μg | |
Primary | |
Detects mouse Cathepsin D in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Cathepsin A, rmCathepsin B, rmCathepsin C, rmCathepsin H, and rmCathepsin X/Z/P is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
Goat | |
Affinity Purified | |
RUO | |
1509 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction