Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cathepsin F Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Cathepsin F |
---|---|
Clone | 192204 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Cathepsin F Monoclonal specifically detects Cathepsin F in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin F | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22 | |
CTSF | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
192204 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
Q9WUT4 | |
8722 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin F Ser20-Asn462 Accession # Q9WUT4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cathepsin F in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse Cathepsin B, C, L, or S is observed and approximately 5% and 25% cross-reactivity with recombinant human Cathepsin F under non-reducing conditions and reducing conditions, respectively, is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title