Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Cathepsin H Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1013
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1013 500 μg
MAB1013SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1013 Supplier R&D Systems Supplier No. MAB1013
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Cathepsin H Monoclonal specifically detects Cathepsin H in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Cathepsin H
Applications Western Blot
Classification Monoclonal
Clone 175909
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q3UCD6
Gene Alias ACC-4, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin H, CPSB, CTSH, DKFZp686B24257, EC 3.4.22, EC 3.4.22.16, MGC1519, minichain, N-benzoylarginine-beta-naphthylamide hydrolase
Gene Symbols CTSH
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin H Glu22-Val333 Accession # Q3UCD6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1512
Test Specificity Detects mouse Cathepsin H in direct ELISAs and Western blots. It recognizes both the pro and active forms of recombinant mouse (rm) Cathepsin H. In Western blots, no cross-reactivity with rmCathepsin A, D, E, X, and Z or recombinant human Cathepsin B, C, L, or S is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.