Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cathepsin X/Z/P Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Supplier: R&D Systems AF1033
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Polyclonal specifically detects Cathepsin X/Z/P in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry.Specifications
Cathepsin X/Z/P | |
Polyclonal | |
Unconjugated | |
Q9WUU7 | |
Ctsz | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin X/Z/P Ala23-Val306 Accession # Q9WUU7 | |
100 μg | |
Primary | |
Detects mouse Cathepsin X/Z/P in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.0 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Cathepsin P, Cathepsin Z, CTSX, CTSZ | |
Goat | |
Affinity Purified | |
RUO | |
1522 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction