Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Ccd1/DIXDC1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5599SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5599SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5599SP Supplier R&D Systems Supplier No. AF5599SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Ccd1/DIXDC1 Polyclonal specifically detects Ccd1/DIXDC1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Ccd1/DIXDC1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q80Y83
Gene Alias CCD1, Coiled-coil protein DIX1, Coiled-coil-DIX1, DIX domain containing 1, DIX domain-containing protein 1, Dixdc1, dixin, Kiaa1735, KIAA1735coiled-coil-DIX1
Gene Symbols DIXDC1
Host Species Goat
Immunogen E. coli-derived recombinant mouse Ccd1/DIXDC1 Gln481-Asn711 Accession # Q80Y83
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85458
Test Specificity Detects mouse Ccd1/DIXDC1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.