Learn More
Mouse CCL1/I-309/TCA-3 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCL1/I-309/TCA-3 |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Clone | 148107 |
| Conjugate | Unconjugated |
| Dilution | Neutralization 0.6-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P10146 |
| Gene Alias | C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3 |
| Gene Symbols | CCL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.