Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL1/I-309/TCA-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB845500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB845500 500 μg
MAB845SP 25 μg
MAB845100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB845500 Supplier R&D Systems Supplier No. MAB845500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

CCL1/I-309/TCA-3 Monoclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL1/I-309/TCA-3
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 148113
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 2-10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10146
Gene Alias C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3
Gene Symbols CCL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CCL1/I-309/TCA-3 Lys24-Cys92 Accession # P10146
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6346
Test Specificity Detects mouse CCL1 in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 23, 24, 25, 26, 27, 28, recombinant mouse CCL2, 3, 4,56, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.