Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL11/Eotaxin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB420500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB420500 500 μg
MAB420SP 25 μg
MAB420100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB420500 Supplier R&D Systems Supplier No. MAB420500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen CCL11/Eotaxin
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 42285
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-30 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P48298
Gene Alias C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin)
Gene Symbols CCL11
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL11/Eotaxin His24-Pro97 Accession # P48298
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6356
Test Specificity Detects mouse CCL11/Eotaxin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CCL11/Eotaxin, recombinant mouse CCL7, and rhCCL21 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.