Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL12/MCP-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4281SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4281SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4281SP Supplier R&D Systems Supplier No. MAB4281SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Neutralization.

Specifications

Antigen CCL12/MCP-5
Applications Neutralization
Classification Monoclonal
Clone 147801
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.5-2.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q62401
Gene Alias C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP5, MCP-5, Scya12
Gene Symbols Ccl12
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL12/MCP-5 Gly23-Gly104 Accession # Q62401
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20293
Test Specificity Detects mouse CCL12/MCP-5 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.